4-(1,2,3-Thiadiazol-4-yl)benzyl bromide

Supplier: Apollo Scientific
Warning

Synonyms: 4-[4-(Bromomethyl)phenyl]-1,2,3-thiadiazole

OR23262-1G
APOSOR23262-1GEA 137 EUR
APOSOR23262-1G
4-(1,2,3-Thiadiazol-4-yl)benzyl bromide
4-(1,2,3-Thiadiazol-4-yl)benzyl bromide
Formula: C₉H₇BrN₂S
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00052102
CAS Number: 163798-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR