(Diphenylphosphoryl)methanol

Supplier: Apollo Scientific
Warning

Synonyms: (Hydroxymethyl)diphenylphosphine oxide, Diphenyl(hydroxymethyl)phosphine oxide

OR25804-250MG
APOSOR25804-250MGEA 98.6 EUR
APOSOR25804-250MG
(Diphenylphosphoryl)methanol
(Diphenylphosphoryl)methanol
Formula: C₁₃H₁₃O₂P
Storage Temperature: Refrigerator
MDL Number: MFCD00087496
CAS Number: 884-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR