2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate 95+%

Supplier: Apollo Scientific
Danger

OR27845-25G OR27845-5G OR27845-1G
APOSOR27845-25GEA 765 EUR
APOSOR27845-25G APOSOR27845-5G APOSOR27845-1G
2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate 95+%
2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate
Formula: C₁₀H₁₁NO₆
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00216510
CAS Number: 205692-61-1
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR