5-Methylpyrazine-2-carboxylic acid

Supplier: Apollo Scientific
Danger

OR28728-100G OR28728-25G OR28728-5G
APOSOR28728-100GEA 39.3 EUR
APOSOR28728-100G APOSOR28728-25G APOSOR28728-5G
5-Methylpyrazine-2-carboxylic acid
5-Methylpyrazine-2-carboxylic acid
Formula: C₆H₆N₂O₂
Melting Pt: 165…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00068241
CAS Number: 5521-55-1
EINECS: 413-260-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR