4,7-Dichloroquinoline

Supplier: Apollo Scientific
Warning

OR28817-100G OR28817-25G
APOSOR28817-100GEA 46.4 EUR
APOSOR28817-100G APOSOR28817-25G
4,7-Dichloroquinoline
4,7-Dichloroquinoline
Formula: C₉H₅Cl₂N
Boiling Pt: 148 °C/10mmHg
Melting Pt: 81…87 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6
EINECS: 201-714-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR