(4-Methylmorpholin-2-yl)methanamine

Supplier: Apollo Scientific

OR300162-1G
APOSOR300162-1GEA 190 EUR
APOSOR300162-1G
(4-Methylmorpholin-2-yl)methanamine
(4-Methylmorpholin-2-yl)methanamine
Formula: C₆H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD03990582
CAS Number: 141814-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR