(2-Methoxypyridin-4-yl)methanol 98%

Supplier: Apollo Scientific

Synonyms: 2-Methoxy-4-pyridinemethanol

OR301205-5G
APOSOR301205-5GEA 109 EUR
APOSOR301205-5G
(2-Methoxypyridin-4-yl)methanol 98%
(2-Methoxypyridin-4-yl)methanol
Formula: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00859298
CAS Number: 123148-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR