1-(2,3-Dihydro-1H-indol-6-yl)ethanone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(Indolin-6-yl)ethanone

OR305614-100MG
APOSOR305614-100MGEA 0 EUR
APOSOR305614-100MG
1-(2,3-Dihydro-1H-indol-6-yl)ethanone
1-(2,3-Dihydro-1H-indol-6-yl)ethanone
Formula: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD18449837
CAS Number: 147265-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR