1H-Pyrrolo[2,3-b]pyridine-2,3-dione

Supplier: Apollo Scientific
Warning

OR305630-250MG
APOSOR305630-250MGEA 116 EUR
APOSOR305630-250MG
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
Formula: C₇H₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09999197
CAS Number: 5654-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR