1-tert-Butyl 2-methyl (2R,4R)-4-aminopyrrolidine-1,2-dicarboxylate 95%

Supplier: Apollo Scientific
Warning

Synonyms: (2R,4R)-4-Amino-1-(tert-butoxycarbonyl)-2-(methoxycarbonyl)pyrrolidine, cis-4-Amino-D-proline methyl ester, N1-BOC protected, Methyl (2R,4R)-4-aminopyrrolidine-2-carboxylate, N1-BOC protected

OR30678-250MG
APOSOR30678-250MGEA 284 EUR
APOSOR30678-250MG
1-tert-Butyl 2-methyl (2R,4R)-4-aminopyrrolidine-1,2-dicarboxylate 95%
1-tert-Butyl 2-methyl (2R,4R)-4-aminopyrrolidine-1,2-dicarboxylate
Formula: C₁₁H₂₀N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD08704544
CAS Number: 1146160-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR