N-(5-Bromopentyl)phthalimide 97%

Supplier: Apollo Scientific

OR3071-25G
APOSOR3071-25GEA 128 EUR
APOSOR3071-25G
N-(5-Bromopentyl)phthalimide 97%
N-(5-Bromopentyl)phthalimide
Formula: C₁₃H₁₄BrNO₂
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00060522
CAS Number: 954-81-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR