3-Piperidin-4-ylmethyl-phenol hydrochloride

Supplier: Apollo Scientific

OR307721-1G
APOSOR307721-1GEA 491 EUR
APOSOR307721-1G
3-Piperidin-4-ylmethyl-phenol hydrochloride
3-Piperidin-4-ylmethyl-phenol hydrochloride
Formula: C₁₂H₁₇NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD08437510
CAS Number: 782504-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR