(R)-4-Boc-(3-hydroxymethyl)morpholine

Supplier: Apollo Scientific

Synonyms: (R)-4-Boc-(3-Hydroxymethyl)morpholine, tert-Butyl (3R)-3-(hydroxymethyl)morpholine-4-carboxylate

OR308156-5G
APOSOR308156-5GEA 133 EUR
APOSOR308156-5G
(R)-4-Boc-(3-hydroxymethyl)morpholine
(R)-4-Boc-(3-hydroxymethyl)morpholine
Formula: C₁₀H₁₉NO₄
MW: 217.2600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06799477
CAS Number: 215917-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR