2-Amino-3-benzyloxypyridine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Benzyloxypyridin-2-amine, 3-Benzyloxy-2-pyridylamine

OR3137-25G OR3137-5G
APOSOR3137-25GEA 40.6 EUR
APOSOR3137-25G APOSOR3137-5G
2-Amino-3-benzyloxypyridine
2-Amino-3-benzyloxypyridine
Formula: C₁₂H₁₂N₂O
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00006316
CAS Number: 24016-03-3
EINECS: 245-983-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR