5-(Chloromethyl)-4-isopropyl-1,2,3-thiadiazole 95+%

Supplier: Apollo Scientific

OR314016-250MG
APOSOR314016-250MGEA 255 EUR
APOSOR314016-250MG
5-(Chloromethyl)-4-isopropyl-1,2,3-thiadiazole 95+%
5-(Chloromethyl)-4-isopropyl-1,2,3-thiadiazole
Formula: C₆H₉ClN₂S
Storage Temperature: Ambient
MDL Number: MFCD11053980
CAS Number: 1060817-54-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR