D(+)-Tryptophan 97%

Supplier: Apollo Scientific

Synonyms: H-D-Trp-OH

OR315019-100G OR315019-500G
APOSOR315019-100GEA 71.2 EUR
APOSOR315019-100G APOSOR315019-500G
D(+)-Tryptophan 97%
D(+)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00005647
CAS Number: 153-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR