tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 98%

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate, 3,8-Diazabicyclo[3.2.1]octane-8-carboxylic acid, 8-BOC protected

OR315163-1G OR315163-250MG OR315163-5G
APOSOR315163-1GEA 40.6 EUR
APOSOR315163-1G APOSOR315163-250MG APOSOR315163-5G
tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 98%
tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate
Formula: C₁₁H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD17016708
CAS Number: 149771-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR