3-Imidazol-1-yl-phenylamine

Supplier: Apollo Scientific
Warning

Synonyms: 3-(1H-Imidazol-1-yl)aniline

OR318066-1G OR318066-10G OR318066-5G
APOSOR318066-1GEA 0 EUR
APOSOR318066-1G APOSOR318066-10G APOSOR318066-5G
3-Imidazol-1-yl-phenylamine
3-Imidazol-1-ylphenylamine
Formula: C₉H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD09258808
CAS Number: 112677-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR