1-(4-Methyl-1,2,5-oxadiazol-3-yl)-1-ethanone 95%

Supplier: Apollo Scientific
Warning

OR33379-1G
APOSOR33379-1GEA 488 EUR
APOSOR33379-1G
1-(4-Methyl-1,2,5-oxadiazol-3-yl)-1-ethanone 95%
1-(4-Methyl-1,2,5-oxadiazol-3-yl)-1-ethanone
Formula: C₅H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00476459
CAS Number: 165067-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR