2-Benzyl-4-methylthiazole-5-carboxylic acid

Supplier: Apollo Scientific

OR346077-1G
APOSOR346077-1GEA 0 EUR
APOSOR346077-1G
2-Benzyl-4-methylthiazole-5-carboxylic acid
2-Benzyl-4-methylthiazole-5-carboxylic acid
Formula: C₁₂H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD07643224
CAS Number: 876715-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR