3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid

Supplier: Apollo Scientific
Warning

OR346226-1G OR346226-5G
APOSOR346226-1GEA 0 EUR
APOSOR346226-1G APOSOR346226-5G
3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid
3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid
Formula: C₁₂H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD11557830
CAS Number: 91569-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR