1,1-Dichloroethane 98%

Supplier: Apollo Scientific
Danger

Synonyms: Ethylidene dichloride

OR3712-5G OR3712-1G OR3712-25G
APOSOR3712-5GEA 197 EUR
APOSOR3712-5G APOSOR3712-1G APOSOR3712-25G
1,1-Dichloroethane 98%
1,1-Dichloroethane
Formula: Cl₂HCCH₃
Boiling Pt: 57 °C
Melting Pt: –98 °C
Density: 1.18 g/cm³ (25 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00013673
CAS Number: 75-34-3
EINECS: 200-863-5
UN: 2362
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR