2-Bromo-4-methoxy-6-nitroaniline

Supplier: Apollo Scientific
Warning

OR400396-1G OR400396-25G OR400396-5G
APOSOR400396-1GEA 26.1 EUR
APOSOR400396-1G APOSOR400396-25G APOSOR400396-5G
2-Bromo-4-methoxy-6-nitroaniline
2-Bromo-4-methoxy-6-nitroaniline
Formula: C₇H₇BrN₂O₃
Melting Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD18088521
CAS Number: 10172-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR