3-Chloro-6-methoxypyridine-2-carbohydrazide

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-6-methoxypicolinohydrazide, 3-Chloro-6-methoxypyridine-2-carboxylic acid hydrazide

OR40368-1G OR40368-250MG
APOSOR40368-1GEA 313 EUR
APOSOR40368-1G APOSOR40368-250MG
3-Chloro-6-methoxypyridine-2-carbohydrazide
3-Chloro-6-methoxypyridine-2-carbohydrazide
Formula: C₇H₈ClN₃O₂
Boiling Pt: 141…142 °C
Storage Temperature: Ambient
MDL Number: MFCD17676113
CAS Number: 1257535-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR