2,4,6-Triisopropylbenzoyl chloride 99%

Supplier: Apollo Scientific
Danger

OR40683-25G OR40683-5G OR40683-1G
APOSOR40683-25GEA 0 EUR
APOSOR40683-25G APOSOR40683-5G APOSOR40683-1G
2,4,6-Triisopropylbenzoyl chloride 99%
2,4,6-Triisopropylbenzoyl chloride
Formula: C₁₆H₂₃ClO
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00015030
CAS Number: 57199-00-5
EINECS: 260-614-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR