4-(2-Methylthiazol-4-yl)phenylamine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(2-Methylthiazol-4-yl)aniline, 4-(2-Methyl-1,3-thiazol-4-yl)aniline

OR4380-250MG OR4380-5G
APOSOR4380-250MGEA 36.4 EUR
APOSOR4380-250MG APOSOR4380-5G
4-(2-Methylthiazol-4-yl)phenylamine 97%
4-(2-Methylthiazol-4-yl)phenylamine
Formula: C₁₀H₁₀N₂S
Storage Temperature: Refrigerator
MDL Number: MFCD02691928
CAS Number: 25021-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR