3-Carboxyphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

OR4442-100G OR4442-25G OR4442-5G
APOSOR4442-100GEA 91.5 EUR
APOSOR4442-100G APOSOR4442-25G APOSOR4442-5G
3-Carboxyphenylboronic acid
3-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
Melting Pt: 240…247 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR