1-Bromo-2-(3-bromopropyl)benzene 97%

Supplier: Apollo Scientific

OR452134-1G
APOSOR452134-1GEA 243 EUR
APOSOR452134-1G
1-Bromo-2-(3-bromopropyl)benzene 97%
1-Bromo-2-(3-bromopropyl)benzene
Formula: C₉H₁₀Br₂
Storage Temperature: Ambient
MDL Number: MFCD09744408
CAS Number: 1075-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR