3-Bromobenzyl chloride

Supplier: Apollo Scientific
Danger

Synonyms: 3-Bromo-α-chlorotoluene, 1-(Chloromethyl)-3-bromobenzene, α-Chloro-3-bromotoluene, 1-Bromo-3-(chloromethyl)benzene

OR4547-25G OR4547-100G
APOSOR4547-25GEA 77 EUR
APOSOR4547-25G APOSOR4547-100G
3-Bromobenzyl chloride
3-Bromobenzyl chloride
Formula: C₇H₆BrCl
Boiling Pt: 118…119 °C/18mmHg
Density: 1.608 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00040865
CAS Number: 932-77-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR