4-Bromo-2-methyl-6-nitroaniline

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

OR4613-5G OR4613-1G OR4613-25G
APOSOR4613-5GEA 159 EUR
APOSOR4613-5G APOSOR4613-1G APOSOR4613-25G
4-Bromo-2-methyl-6-nitroaniline
4-Bromo-2-methyl-6-nitroaniline
Formula: C₇H₇BrN₂O₂
Melting Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00052919
CAS Number: 77811-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR