2,6-Dibromo-4-methylpyridine

Supplier: Apollo Scientific
Warning

OR46507-10G OR46507-1G OR46507-5G
APOSOR46507-10GEA 316 EUR
APOSOR46507-10G APOSOR46507-1G APOSOR46507-5G
2,6-Dibromo-4-methylpyridine
2,6-Dibromo-4-methylpyridine
Formula: C₆H₅Br₂N
Melting Pt: 74…79 °C
Storage Temperature: Ambient
MDL Number: MFCD11036225
CAS Number: 73112-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR