(R,R)-(-)-2,5-Hexanediol

Supplier: Apollo Scientific
Warning

Synonyms: (2R,5R)-2,5-Hexanediol, (R,R)-(-)-2,5-Dihydroxy hexane, (2R,5R)-Hexane-2,5-diol

OR46574-1G OR46574-5G
APOSOR46574-1GEA 68.3 EUR
APOSOR46574-1G APOSOR46574-5G
(R,R)-(-)-2,5-Hexanediol
(R,R)-(-)-2,5-Hexanediol
Formula: C₆H₁₄O₂
Melting Pt: 52…54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142339
CAS Number: 17299-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR