3-(2-Pyridyl)propanamide

Supplier: Apollo Scientific

OR470610-5G
APOSOR470610-5GEA 997 EUR
APOSOR470610-5G
3-(2-Pyridyl)propanamide
3-(2-Pyridyl)propanamide
Formula: C₈H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD18384619
CAS Number: 84199-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR