1-(1-Methyl-1H-indol-3-yl)ethanone

Supplier: Apollo Scientific

Synonyms: 3-Acetyl-1-methylindole

OR471162-1G
APOSOR471162-1GEA 397 EUR
APOSOR471162-1G
1-(1-Methyl-1H-indol-3-yl)ethanone
1-(1-Methyl-1H-indol-3-yl)ethanone
Formula: C₁₁H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00540232
CAS Number: 19012-02-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR