4-Chloro-6-(methylamino)pyrimidine-5-carbonitrile

Supplier: Apollo Scientific

OR471391-1G
APOSOR471391-1GEA 916 EUR
APOSOR471391-1G
4-Chloro-6-(methylamino)pyrimidine-5-carbonitrile
4-Chloro-6-(methylamino)pyrimidine-5-carbonitrile
Formula: C₆H₅ClN₄
Storage Temperature: Ambient
MDL Number: MFCD07761813
CAS Number: 872890-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR