1-(Hydroxymethyl)cyclopropanecarboxamide 95%

Supplier: Apollo Scientific

OR480277-10G OR480277-1G OR480277-5G
APOSOR480277-10GEA 1280 EUR
APOSOR480277-10G APOSOR480277-1G APOSOR480277-5G
1-(Hydroxymethyl)cyclopropanecarboxamide 95%
1-(Hydroxymethyl)cyclopropanecarboxamide
Formula: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12198439
CAS Number: 1123169-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR