3,5-Dimethylbenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR4988-100G OR4988-1KG OR4988-500G
APOSOR4988-100GEA 21.9 EUR
APOSOR4988-100G APOSOR4988-1KG APOSOR4988-500G
3,5-Dimethylbenzoic acid 98%
3,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR