Benzeneruthenium(II) chloride dimer 95+%

Supplier: Apollo Scientific
Warning

Synonyms: Dichloro(benzene)ruthenium(II) dimer

OR54465-1G OR54465-5G
APOSOR54465-1GEA 55.1 EUR
APOSOR54465-1G APOSOR54465-5G
Benzeneruthenium(II) chloride dimer 95+%
Benzeneruthenium(II) chloride dimer
Formula: C₁₂H₁₂Cl₄Ru₂
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064686
CAS Number: 37366-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR