(S)-(+)-2-Methylpiperazine 97%

Supplier: Apollo Scientific
Danger

Synonyms: (S)-2-Methylpiperazine

OR5877-10G
APOSOR5877-10GEA 21.9 EUR
APOSOR5877-10G
(S)-(+)-2-Methylpiperazine 97%
(S)-(+)-2-Methylpiperazine

Chiral intermediate for pharmaceuticals and agrochemicals.

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Melting Pt: 89…91 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00171452
CAS Number: 74879-18-8
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR