O,O-Diethyl S-[(ethylsulphinyl)methyl] thiophosphate solution 100 ng/µl in cyclohexane

Supplier: Apollo Scientific
Danger

Synonyms: Phorate-oxon-sulphoxide, O,O-Diethyl S-[(ethylsulphinyl)methyl] phosphorothioate

OR59940-1ML
APOSOR59940-1MLEA 281 EUR
APOSOR59940-1ML
O,O-Diethyl S-[(ethylsulphinyl)methyl] thiophosphate solution 100 ng/µl in cyclohexane
O,O-Diethyl S-[(ethylsulphinyl)methyl] thiophosphate
Formula: C₇H₁₇O₄PS₂
Boiling Pt: 80.7 °C
Melting Pt: 6.5 °C
Density: 0.78 g/cm³ (25 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00155406
CAS Number: 2588-05-8
UN: 1145
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR