2-Methylsulphonyl-1-(pyridin-2-yl)ethanone 95+%

Supplier: Apollo Scientific

OR7991-1G OR7991-5G
APOSOR7991-1GEA 130 EUR
APOSOR7991-1G APOSOR7991-5G
2-Methylsulphonyl-1-(pyridin-2-yl)ethanone 95+%
2-Methylsulphonyl-1-(pyridin-2-yl)ethanone
Formula: C₈H₉NO₃S
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD01941305
CAS Number: 27302-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR