2,5-Dichlorothiophenol 98%

Supplier: Apollo Scientific
Warning

OR7999-100G OR7999-25G OR7999-5G
APOSOR7999-100GEA 1330 EUR
APOSOR7999-100G APOSOR7999-25G APOSOR7999-5G
2,5-Dichlorothiophenol 98%
2,5-Dichlorothiophenol
Formula: C₆H₄Cl₂S
Boiling Pt: 112…116 °C/50mmHg
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004831
CAS Number: 5858-18-4
EINECS: 227-486-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR