Methyl bromopyruvate 90%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3-Bromopyruvate, Methyl 3-Bromo-2-oxopropionate, Bromopyruvic acid methyl ester, 3-Bromopyruvic acid methyll ester, 3-Bromo-2-oxopropionic acid methyll ester, 3-Bromo-2-oxopropionic acid methyl ester

OR8931-25G
APOSOR8931-25GEA 109 EUR
APOSOR8931-25G
Methyl bromopyruvate 90%
Methyl bromopyruvate
Formula: BrH₂CC(O)COOCH₃
Density: 1.7 g/cm³ (25 °C)
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862963
CAS Number: 7425-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR