(R)-Ethyl-2-hydroxypropanoate 95%

Supplier: Apollo Scientific

Synonyms: (+)-Ethyl d-lactate, (R)-Ethyl 2-hydroxypropanoate

OR902275-100G OR902275-25G OR902275-5G
APOSOR902275-100GEA 53.8 EUR
APOSOR902275-100G APOSOR902275-25G APOSOR902275-5G
(R)-Ethyl-2-hydroxypropanoate 95%
(R)-Ethyl-2-hydroxypropanoate
Formula: C₅H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00077825
CAS Number: 7699-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR