5,6-Dichloro-2,3-dicyanopyrazine 96%

Supplier: Apollo Scientific

Synonyms: 5,6-Dichloropyrazine-2,3-dicarbonitrile, 5,6-Dichloro-2,3-pyrazinedicarbonitrile

OR902743-25G
APOSOR902743-25GEA 221 EUR
APOSOR902743-25G
5,6-Dichloro-2,3-dicyanopyrazine 96%
5,6-Dichloro-2,3-dicyanopyrazine
Formula: C₆Cl₂N₄
Storage Temperature: Ambient
MDL Number: MFCD00191419
CAS Number: 56413-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR