Methyl-3,5-dimethylbenzoate 97%

Supplier: Apollo Scientific

Synonyms: Methyl 3,5-dimethylbenzoate

OR907873-100G
APOSOR907873-100GEA 134 EUR
APOSOR907873-100G
Methyl-3,5-dimethylbenzoate 97%
Methyl 3,5-dimethylbenzoate
Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00077971
CAS Number: 25081-39-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR