(6-Chloro-pyridazin-3-yl)dimethylamine

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-N,N-dimethylpyridazin-3-amine

OR909507-1G OR909507-5G
APOSOR909507-1GEA 34.8 EUR
APOSOR909507-1G APOSOR909507-5G
(6-Chloro-pyridazin-3-yl)dimethylamine
(6-Chloro-pyridazin-3-yl)dimethylamine
Formula: C₆H₈ClN₃
Melting Pt: 108…109 °C
Storage Temperature: Ambient
MDL Number: MFCD02732844
CAS Number: 7145-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR