tert-Butyl N-(2-chloroethyl)-N-methylcarbamate 96%

Supplier: Apollo Scientific
Danger

Synonyms: tert-Butyl (2-chloroethyl)(methyl)carbamate

OR909760-1G OR909760-5G OR909760-10G
APOSOR909760-1GEA 132 EUR
APOSOR909760-1G APOSOR909760-5G APOSOR909760-10G
tert-Butyl N-(2-chloroethyl)-N-methylcarbamate 96%
tert-Butyl N-(2-chloroethyl)-N-methylcarbamate
Formula: C₈H₁₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD20624610
CAS Number: 220074-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR