2,4-Dimethylbenzaldehyde 95%

Supplier: Apollo Scientific

OR911450-100G OR911450-25G
APOSOR911450-100GEA 456 EUR
APOSOR911450-100G APOSOR911450-25G
2,4-Dimethylbenzaldehyde 95%
2,4-Dimethylbenzaldehyde
Formula: C₉H₁₀O
Storage Temperature: Ambient
MDL Number: MFCD00003340
CAS Number: 15764-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR