3-(Chloromethyl)-5-phenylisoxazole 97%

Supplier: Apollo Scientific
Danger

OR9185-1G
APOSOR9185-1GEA 118 EUR
APOSOR9185-1G
3-(Chloromethyl)-5-phenylisoxazole 97%
3-(Chloromethyl)-5-phenylisoxazole
Formula: C₁₀H₈ClNO
Melting Pt: 48…49 °C
Storage Temperature: Ambient
MDL Number: MFCD01444154
CAS Number: 14731-10-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR